Anti-ERBB3 Mouse Monoclonal Antibody [clone: 13B268]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030190EA 3080 DKK
USBI030190
Anti-ERBB3 Mouse Monoclonal Antibody [clone: 13B268]
Antistoffer
Type: Primary
Antigen: ERBB3 (v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 3)
Clonality: Monoclonal
Clone: 13B268
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR