Anti-ERBB2 Mouse Monoclonal Antibody [clone: 13B266]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030188EA 3080 DKK
USBI030188
Anti-ERBB2 Mouse Monoclonal Antibody [clone: 13B266]
Antistoffer
Type: Primary
Antigen: ERBB2 (v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 2)
Clonality: Monoclonal
Clone: 13B266
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR