Anti-EPHB4 Mouse Monoclonal Antibody [clone: 13B260]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030182EA 3930 DKK
USBI030182
Anti-EPHB4 Mouse Monoclonal Antibody [clone: 13B260]
Antistoffer
Type: Primary
Antigen: EPHB4 (EPH receptor B4)
Clonality: Monoclonal
Clone: 13B260
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR