Anti-EPHB3 Mouse Monoclonal Antibody [clone: 13B257]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030179EA 4230 DKK
USBI030179
Anti-EPHB3 Mouse Monoclonal Antibody [clone: 13B257]
Antistoffer
Type: Primary
Antigen: EPHB3 (EPH receptor B3)
Clonality: Monoclonal
Clone: 13B257
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR