Anti-ELK1 Mouse Monoclonal Antibody [clone: 13B243]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030165EA 2840 DKK
USBI030165
Anti-ELK1 Mouse Monoclonal Antibody [clone: 13B243]
Antistoffer
Type: Primary
Antigen: ELK1 (ELK1, member of ETS oncogene family)
Clonality: Monoclonal
Clone: 13B243
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR