Anti-EIF4E Mouse Monoclonal Antibody [clone: 13B241]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030163EA 4410 DKK
USBI030163
Anti-EIF4E Mouse Monoclonal Antibody [clone: 13B241]
Antistoffer
Type: Primary
Antigen: EIF4E (eukaryotic translation initiation factor 4E)
Clonality: Monoclonal
Clone: 13B241
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR