Anti-Delta 4 Mouse Monoclonal Antibody [clone: 13B222]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030144EA 3270 DKK
USBI030144
Anti-Delta 4 Mouse Monoclonal Antibody [clone: 13B222]
Antistoffer
Type: Primary
Antigen: Delta 4
Clonality: Monoclonal
Clone: 13B222
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b+IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR