Anti-CRTC1 Mouse Monoclonal Antibody [clone: 13B194]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030116EA 3080 DKK
USBI030116
Anti-CRTC1 Mouse Monoclonal Antibody [clone: 13B194]
Antistoffer
Type: Primary
Antigen: CRTC1 (CREB regulated transcription coactivator 1)
Clonality: Monoclonal
Clone: 13B194
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR