Anti-CREB1 Mouse Monoclonal Antibody [clone: 13B189]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030111EA 4410 DKK
USBI030111
Anti-CREB1 Mouse Monoclonal Antibody [clone: 13B189]
Antistoffer
Type: Primary
Antigen: CREB1 (cAMP responsive element binding protein 1)
Clonality: Monoclonal
Clone: 13B189
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR