Anti-KRT17 Mouse Monoclonal Antibody [clone: 13B180]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030102EA 3180 DKK
USBI030102
Anti-KRT17 Mouse Monoclonal Antibody [clone: 13B180]
Antistoffer
Type: Primary
Antigen: KRT17 (keratin 17)
Clonality: Monoclonal
Clone: 13B180
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR