Anti-CHUK Mouse Monoclonal Antibody [clone: 13B175]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030097EA 2840 DKK
USBI030097
Anti-CHUK Mouse Monoclonal Antibody [clone: 13B175]
Antistoffer
Type: Primary
Antigen: CHUK (conserved helix-loop-helix ubiquitous kinase)
Clonality: Monoclonal
Clone: 13B175
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR