Anti-CER1 Mouse Monoclonal Antibody [clone: 13B170]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030092EA 3080 DKK
USBI030092
Anti-CER1 Mouse Monoclonal Antibody [clone: 13B170]
Antistoffer
Type: Primary
Antigen: CER1 (cerberus 1, DAN family BMP antagonist)
Clonality: Monoclonal
Clone: 13B170
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR