Anti-CDKN1C Mouse Monoclonal Antibody [clone: 13B162]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030084EA 3080 DKK
USBI030084
Anti-CDKN1C Mouse Monoclonal Antibody [clone: 13B162]
Antistoffer
Type: Primary
Antigen: CDKN1C (cyclin-dependent kinase inhibitor 1C (p57, Kip2))
Clonality: Monoclonal
Clone: 13B162
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR