Anti-CDK9 Mouse Monoclonal Antibody [clone: 13B160]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030082EA 4410 DKK
USBI030082
Anti-CDK9 Mouse Monoclonal Antibody [clone: 13B160]
Antistoffer
Type: Primary
Antigen: CDK9 (cyclin-dependent kinase 9)
Clonality: Monoclonal
Clone: 13B160
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR