Anti-CDH1 Mouse Monoclonal Antibody [clone: 13B153]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030075EA 3080 DKK
USBI030075
Anti-CDH1 Mouse Monoclonal Antibody [clone: 13B153]
Antistoffer
Type: Primary
Antigen: CDH1 (cadherin 1, type 1, E-cadherin (epithelial))
Clonality: Monoclonal
Clone: 13B153
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR