Anti-CDC27 Mouse Monoclonal Antibody [clone: 13B151]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030073EA 3080 DKK
USBI030073
Anti-CDC27 Mouse Monoclonal Antibody [clone: 13B151]
Antistoffer
Type: Primary
Antigen: CDC27 (cell division cycle 27)
Clonality: Monoclonal
Clone: 13B151
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR