Anti-CD80 Mouse Monoclonal Antibody [clone: 13B143]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030065EA 3080 DKK
USBI030065
Anti-CD80 Mouse Monoclonal Antibody [clone: 13B143]
Antistoffer
Type: Primary
Antigen: CD80 (CD80 molecule)
Clonality: Monoclonal
Clone: 13B143
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR