Anti-CD4 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 13B133]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030055EA 2770 DKK
USBI030055
Anti-CD4 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 13B133]
Antistoffer
Type: Primary
Antigen: CD4 (CD4 molecule)
Clonality: Monoclonal
Clone: 13B133
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR