Anti-CD34 Mouse Monoclonal Antibody [clone: 13B128]
Leverandør: United States Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBI030050EA 4230 DKK
USBI030050
Anti-CD34 Mouse Monoclonal Antibody [clone: 13B128]
Antistoffer
Recommended dilution: Western Blot: 1:500-1:2000; Immunochemistry: 1:200-1:1000; Immunofluorescence: 1:200-1:1000; ELISA: 1:10000
Type: Primary
Antigen: CD34 (CD34 molecule)
Clonality: Monoclonal
Clone: 13B128
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human