Anti-CD3 Mouse Monoclonal Antibody [clone: 13B123]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030045EA 2350 DKK
USBI030045
Anti-CD3 Mouse Monoclonal Antibody [clone: 13B123]
Antistoffer
Type: Primary
Antigen: CD3
Clonality: Monoclonal
Clone: 13B123
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR