Anti-ITGB2 Mouse Monoclonal Antibody [clone: 13B115]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030037EA 3080 DKK
USBI030037
Anti-ITGB2 Mouse Monoclonal Antibody [clone: 13B115]
Antistoffer
Type: Primary
Antigen: ITGB2 (integrin, beta 2 (complement component 3 receptor 3 and 4 subunit))
Clonality: Monoclonal
Clone: 13B115
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR