Anti-CD14 Mouse Monoclonal Antibody [clone: 13B113]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030035EA 2840 DKK
USBI030035
Anti-CD14 Mouse Monoclonal Antibody [clone: 13B113]
Antistoffer
Type: Primary
Antigen: CD14 (CD14 molecule)
Clonality: Monoclonal
Clone: 13B113
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR