Anti-CASP8 Mouse Monoclonal Antibody [clone: 13B100]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030022EA 3270 DKK
USBI030022
Anti-CASP8 Mouse Monoclonal Antibody [clone: 13B100]
Antistoffer
Type: Primary
Antigen: CASP8 (caspase 8, apoptosis-related cysteine peptidase)
Clonality: Monoclonal
Clone: 13B100
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR