Anti-CARM1 Mouse Monoclonal Antibody [clone: 13B99]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030021EA 3270 DKK
USBI030021
Anti-CARM1 Mouse Monoclonal Antibody [clone: 13B99]
Antistoffer
Type: Primary
Antigen: CARM1 (coactivator-associated arginine methyltransferase 1)
Clonality: Monoclonal
Clone: 13B99
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR