Anti-CALR Mouse Monoclonal Antibody [clone: 13B96]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030018EA 3330 DKK
USBI030018
Anti-CALR Mouse Monoclonal Antibody [clone: 13B96]
Antistoffer
Type: Primary
Antigen: CALR (calreticulin)
Clonality: Monoclonal
Clone: 13B96
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR