Anti-CA9 Mouse Monoclonal Antibody [clone: 13B92]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030014EA 3930 DKK
USBI030014
Anti-CA9 Mouse Monoclonal Antibody [clone: 13B92]
Antistoffer
Recommended dilution: Western Blot: 1:500-1:2000; Immunochemistry: 1:200-1:1000; Flow Cytometry: 1:200-1:400; ELISA: 1:10000

Type: Primary
Antigen: CA9 (carbonic anhydrase IX)
Clonality: Monoclonal
Clone: 13B92
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR