Anti-JUN Mouse Monoclonal Antibody [clone: 13B87]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030009EA 3270 DKK
USBI030009
Anti-JUN Mouse Monoclonal Antibody [clone: 13B87]
Antistoffer
Type: Primary
Antigen: JUN (jun proto-oncogene)
Clonality: Monoclonal
Clone: 13B87
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR