Anti-CBL Mouse Monoclonal Antibody [clone: 13B86]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030008EA 3270 DKK
USBI030008
Anti-CBL Mouse Monoclonal Antibody [clone: 13B86]
Antistoffer
Type: Primary
Antigen: CBL (Cbl proto-oncogene, E3 ubiquitin protein ligase)
Clonality: Monoclonal
Clone: 13B86
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR