Anti-BMPR2 Mouse Monoclonal Antibody [clone: 13B75]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI029997EA 3270 DKK
USBI029997
Anti-BMPR2 Mouse Monoclonal Antibody [clone: 13B75]
Antistoffer
Type: Primary
Antigen: BMPR2 (bone morphogenetic protein receptor, type II (serine/threonine kinase))
Clonality: Monoclonal
Clone: 13B75
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR