Anti-BMP4 Mouse Monoclonal Antibody [clone: 13B74]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI029996EA 3080 DKK
USBI029996
Anti-BMP4 Mouse Monoclonal Antibody [clone: 13B74]
Antistoffer
Type: Primary
Antigen: BMP4 (bone morphogenetic protein 4)
Clonality: Monoclonal
Clone: 13B74
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR