Anti-BID Mouse Monoclonal Antibody [clone: 13B66]
Leverandør: United States Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBI029988EA 4410 DKK
USBI029988
Anti-BID Mouse Monoclonal Antibody [clone: 13B66]
Antistoffer
Type: Primary
Antigen: BID (BH3 interacting domain death agonist)
Clonality: Monoclonal
Clone: 13B66
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human