Anti-ATM Mouse Monoclonal Antibody [clone: 13B52]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI029974EA 3270 DKK
USBI029974
Anti-ATM Mouse Monoclonal Antibody [clone: 13B52]
Antistoffer
Type: Primary
Antigen: ATM (ataxia telangiectasia mutated)
Clonality: Monoclonal
Clone: 13B52
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR