Anti-APOB Mouse Monoclonal Antibody [clone: 13B39]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI029961EA 3270 DKK
USBI029961
Anti-APOB Mouse Monoclonal Antibody [clone: 13B39]
Antistoffer
Type: Primary
Antigen: APOB (apolipoprotein B)
Clonality: Monoclonal
Clone: 13B39
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR