Anti-APOA5 Mouse Monoclonal Antibody [clone: 13B37]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI029959EA 3570 DKK
USBI029959
Anti-APOA5 Mouse Monoclonal Antibody [clone: 13B37]
Antistoffer
Type: Primary
Antigen: APOA5 (apolipoprotein A-V)
Clonality: Monoclonal
Clone: 13B37
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR