Anti-AMACR Mouse Monoclonal Antibody [clone: 13B30]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI029952EA 3550 DKK
USBI029952
Anti-AMACR Mouse Monoclonal Antibody [clone: 13B30]
Antistoffer
Type: Primary
Antigen: AMACR (alpha-methylacyl-CoA racemase)
Clonality: Monoclonal
Clone: 13B30
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR