Anti-Serum Albumin Mouse Monoclonal Antibody [clone: 13B23]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI029945EA 3180 DKK
USBI029945
Anti-Serum Albumin Mouse Monoclonal Antibody [clone: 13B23]
Antistoffer
Type: Primary
Antigen: Serum Albumin
Clonality: Monoclonal
Clone: 13B23
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR