Anti-ACTA2 Mouse Monoclonal Antibody [clone: 13B13]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI029935EA 4410 DKK
USBI029935
Anti-ACTA2 Mouse Monoclonal Antibody [clone: 13B13]
Antistoffer
Type: Primary
Antigen: ACTA2 (actin, alpha 2, smooth muscle, aorta)
Clonality: Monoclonal
Clone: 13B13
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR