Anti-ACTA2 Mouse Monoclonal Antibody [clone: 13B12]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI029934EA 3270 DKK
USBI029934
Anti-ACTA2 Mouse Monoclonal Antibody [clone: 13B12]
Antistoffer
Type: Primary
Antigen: ACTA2 (actin, alpha 2, smooth muscle, aorta)
Clonality: Monoclonal
Clone: 13B12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR