Anti-SERPINA1 Mouse Monoclonal Antibody [clone: 13B5]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI029927EA 3080 DKK
USBI029927
Anti-SERPINA1 Mouse Monoclonal Antibody [clone: 13B5]
Antistoffer
Type: Primary
Antigen: SERPINA1 (serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 1)
Clonality: Monoclonal
Clone: 13B5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR