Anti-A1BG Mouse Monoclonal Antibody [clone: 13B4]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI029926EA 3080 DKK
USBI029926
Anti-A1BG Mouse Monoclonal Antibody [clone: 13B4]
Antistoffer
Type: Primary
Antigen: A1BG (alpha-1-B glycoprotein)
Clonality: Monoclonal
Clone: 13B4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR