Anti-KITLG Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI029882EA 3360 DKK
USBI029882
Anti-KITLG Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: ELISA: 1-4?g/ml; Western Blot: 0.5-1 ?g/ml; Neutralization: 0.1-2?g/ml

Type: Primary
Antigen: KITLG (KIT ligand)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR