Anti-TNFRSF11B Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI029857EA 3070 DKK
USBI029857
Anti-TNFRSF11B Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot: 0.5-4?g/ml

Type: Primary
Antigen: TNFRSF11B (tumor necrosis factor receptor superfamily, member 11b)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR