Anti-IL12B Goat Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI029809EA 3210 DKK
USBI029809
Anti-IL12B Goat Polyclonal Antibody
Antistoffer
Recommended dilution: ELISA: 1-4?g/ml; Western Blot: 0.5-1 ?g/ml; Neutralization: 0.1-0.5 ?g/ml

Type: Primary
Antigen: IL12B (interleukin 12B (natural killer cell stimulatory factor 2, cytotoxic lymphocyte maturation factor 2, p40))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR