Anti-HA tag Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 13A235]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI029799EA 3350 DKK
USBI029799
Anti-HA tag Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 13A235]
Antistoffer
Recommended dilution: Immunofluorescence: 2-10 ?g/ml

Type: Tag
Antigen: HA tag (HA tag)
Clonality: Monoclonal
Clone: 13A235
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR