Anti-GFP Mouse Monoclonal Antibody [clone: 13A232]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI029791EA 3530 DKK
USBI029791
Anti-GFP Mouse Monoclonal Antibody [clone: 13A232]
Antistoffer
Recommended dilution: Immunofluorescence: 10-20 ?g/ml; Western Blot: 0.5-4?g/ml; Immunoprecipitation: 2-8?g/ml; Immunochemistry: 10-20 ?g/ml

Type: Tag
Antigen: GFP (green fluorescent protein)
Clonality: Monoclonal
Clone: 13A232
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR