Anti-CSF2 Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI029790EA 3360 DKK
USBI029790
Anti-CSF2 Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: ELISA: 2-3?g/ml; Western Blot: 0.5-4?g/ml; Neutralization: 5-10 ?g/ml

Type: Primary
Antigen: CSF2 (colony stimulating factor 2 (granulocyte-macrophage))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR