Anti-CTGF Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI029757EA 3210 DKK
USBI029757
Anti-CTGF Rabbit Polyclonal Antibody
Antistoffer
  • Recommended dilution: Western Blot: 0.5-4μg/ml; Immunochemistry: 10-20 μg/ml

Type: Primary
Antigen: CTGF (connective tissue growth factor)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR