Anti-BMP2 Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI029736EA 3150 DKK
USBI029736
Anti-BMP2 Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot: 0.5-2?g/ml

Type: Primary
Antigen: BMP2 (bone morphogenetic protein 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR