Anti-BFP Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI029729EA 3550 DKK
USBI029729
Anti-BFP Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot: 0.5-4?g/ml; Immunoprecipitation: 10-20 ?g/ml

Type: Tag
Antigen: BFP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR