Anti-APP Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI029722EA 3210 DKK
USBI029722
Anti-APP Rabbit Polyclonal Antibody
Antistoffer
  • Recommended dilution: Western Blot: 1-4μg/ml; Immunochemistry: 2-10 μg/ml

Type: Primary
Antigen: APP (amyloid beta (A4) precursor protein)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Chicken, Human, Monkey, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR